Assay Details
| Gene Symbol: CG4945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299563.1 | NP_001286492.1 | 6 - 7 | 1409 | 75 | |
| NM_166182.3 | NP_725590.1 | 6 - 7 | 1508 | 75 | ||
| NM_176202.3 | NP_788382.1 | 6 - 7 | 1493 | 75 | ||
| NM_176203.3 | NP_788383.1 | 6 - 7 | 1657 | 75 | ||
| GenBank mRNA | AY070513.1 | - | 6 - 7 | 1484 | 75 | |
More Information
Set Membership: |
|

CG4945
Genomic Map