Assay Details
| Gene Symbol: CG4945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166182.3 | NP_725590.1 | 7 - 8 | 1878 | 76 | |
| GenBank mRNA | AY070513.1 | - | 7 - 8 | 1854 | 76 | |
More Information
Set Membership: |
|

CG4945
Genomic Map