Assay Details
| Gene Symbol: Pkc53E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103880.3 | NP_001097350.1 | 13 - 14 | 2114 | 75 | |
| NM_001259448.1 | NP_001246377.1 | 8 - 9 | 1445 | 75 | ||
| NM_001259449.1 | NP_001246378.1 | 13 - 14 | 2054 | 75 | ||
| NM_001274106.1 | NP_001261035.1 | 12 - 13 | 2113 | 75 | ||
| NM_057334.3 | NP_476682.1 | 12 - 13 | 2596 | 75 | ||
| NM_166201.3 | NP_725626.1 | 13 - 14 | 2117 | 75 | ||
| GenBank mRNA | AY095003.1 | - | 13 - 14 | 2088 | 75 | |
| BT120171.1 | - | 8 - 9 | 1445 | 75 | ||
More Information
Set Membership: |
|

Pkc53E
Genomic Map