Assay Details
| Gene Symbol: Pkc53E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274106.1 | NP_001261035.1 | 5 - 6 | 821 | 67 | |
| NM_057334.3 | NP_476682.1 | 5 - 6 | 1304 | 67 | ||
| NM_166201.3 | NP_725626.1 | 6 - 7 | 825 | 67 | ||
| GenBank mRNA | AY095003.1 | - | 6 - 7 | 796 | 67 | |
More Information
Set Membership: |
|

Pkc53E
Genomic Map