Assay Details
| Gene Symbol: NT5E-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137374.3 | NP_611218.1 | 2 - 3 | 357 | 90 | |
| NM_166232.2 | NP_725681.1 | 2 - 3 | 255 | 90 | ||
| GenBank mRNA | AY069711.1 | - | 2 - 3 | 334 | 90 | |
More Information
Set Membership: |
|

NT5E-2
Genomic Map