Assay Details
| Gene Symbol: NT5E-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137374.3 | NP_611218.1 | 4 - 5 | 759 | 73 | |
| NM_166232.2 | NP_725681.1 | 4 - 5 | 657 | 73 | ||
| GenBank mRNA | AY069711.1 | - | 4 - 5 | 736 | 73 | |
More Information
Set Membership: |
|

NT5E-2
Genomic Map