Assay Details
| Gene Symbol: grh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259472.1 | NP_001246401.1 | 3 - 4 | 989 | 63 | |
| NM_057496.5 | NP_476844.2 | 5 - 6 | 3295 | 63 | ||
| NM_057497.4 | NP_476845.2 | 5 - 6 | 3916 | 63 | ||
| NM_166252.3 | NP_725725.1 | 3 - 4 | 983 | 63 | ||
More Information
Set Membership: |
|

grh
Genomic Map