Assay Details
| Gene Symbol: grh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259472.1 | NP_001246401.1 | 4 - 5 | 1227 | 76 | |
| NM_001299622.1 | NP_001286551.1 | 5 - 6 | 4185 | 76 | ||
| NM_057494.4 | NP_476842.2 | 4 - 5 | 2723 | 76 | ||
| NM_057495.4 | NP_476843.2 | 4 - 5 | 2723 | 76 | ||
| NM_057496.5 | NP_476844.2 | 6 - 7 | 3533 | 76 | ||
| NM_057497.4 | NP_476845.2 | 6 - 7 | 4154 | 76 | ||
| NM_166250.4 | NP_725723.2 | 4 - 5 | 1393 | 76 | ||
| NM_166252.3 | NP_725725.1 | 4 - 5 | 1221 | 76 | ||
| GenBank mRNA | BT029431.1 | - | 4 - 5 | 2417 | 76 | |
More Information
Set Membership: |
|

grh
Genomic Map