Assay Details
| Gene Symbol: grh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259472.1 | NP_001246401.1 | 7 - 8 | 1589 | 66 | |
| NM_001299622.1 | NP_001286551.1 | 8 - 9 | 4547 | 66 | ||
| NM_057494.4 | NP_476842.2 | 7 - 8 | 3085 | 66 | ||
| NM_057495.4 | NP_476843.2 | 7 - 8 | 3085 | 66 | ||
| NM_057496.5 | NP_476844.2 | 9 - 10 | 3895 | 66 | ||
| NM_057497.4 | NP_476845.2 | 9 - 10 | 4516 | 66 | ||
| NM_166250.4 | NP_725723.2 | 7 - 8 | 1755 | 66 | ||
| NM_166252.3 | NP_725725.1 | 7 - 8 | 1583 | 66 | ||
| GenBank mRNA | BT029431.1 | - | 7 - 8 | 2779 | 66 | |
More Information
Set Membership: |
|

grh
Genomic Map