Assay Details
| Gene Symbol: Ntan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169728.2 | NP_001163199.1 | 4 - 5 | 899 | 71 | |
| NM_001169729.2 | NP_001163200.1 | 4 - 5 | 1005 | 71 | ||
| NM_137511.4 | NP_611355.2 | 4 - 5 | 895 | 71 | ||
| NM_166300.3 | NP_725811.1 | 4 - 5 | 1005 | 71 | ||
| GenBank mRNA | AF239612.1 | - | 4 - 5 | 841 | 71 | |
| AY118895.1 | - | 4 - 5 | 874 | 71 | ||
More Information
Set Membership: |
|

Ntan1
Genomic Map