Assay Details
| Gene Symbol: 5-HT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144236.2 | NP_001137708.2 | 2 | 651 | 58 | |
| NM_001169730.3 | NP_001163201.2 | 2 | 651 | 58 | ||
| NM_079065.6 | NP_523789.3 | 2 | 651 | 58 | ||
| GenBank mRNA | BT072928.1 | - | 2 | 632 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT072928.1, NM_001144236.2, NM_001169730.3, NM_079065.6 transcripts.
Set Membership: |
|

5-HT1B
Genomic Map