Assay Details
| Gene Symbol: hppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299675.1 | NP_001286604.1 | 3 - 4 | 919 | 114 | |
| NM_001299676.1 | NP_001286605.1 | 3 - 4 | 919 | 114 | ||
| NM_001299677.1 | NP_001286606.1 | 3 - 4 | 919 | 114 | ||
| NM_137555.3 | NP_611399.2 | 3 - 4 | 919 | 114 | ||
| NM_166335.3 | NP_725863.1 | 3 - 4 | 919 | 114 | ||
| GenBank mRNA | BT150295.1 | - | 3 - 4 | 903 | 114 | |
More Information
Set Membership: |
|

hppy
Genomic Map