Assay Details
| Gene Symbol: Fak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144246.3 | NP_001137718.1 | 6 - 7 | 2394 | 78 | |
| NM_001144247.3 | NP_001137719.2 | 7 - 8 | 1823 | 78 | ||
| NM_001169736.2 | NP_001163207.1 | 6 - 7 | 2394 | 78 | ||
| NM_079069.4 | NP_523793.2 | 6 - 7 | 2394 | 78 | ||
| NM_166352.3 | NP_725891.1 | 7 - 8 | 2180 | 78 | ||
| NM_166353.3 | NP_725892.1 | 7 - 8 | 1921 | 78 | ||
| GenBank mRNA | BT133413.1 | - | 6 - 7 | 2377 | 78 | |
More Information
Set Membership: |
|

Fak
Genomic Map