Assay Details
| Gene Symbol: Obp57a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166395.3 | NP_725966.1 | 1 - 2 | 78 | 150 | |
| GenBank mRNA | AF457148.1 | - | 1 - 2 | 59 | 150 | |
| BT023329.1 | - | 1 - 2 | 61 | 150 | ||
More Information
Set Membership: |
|

Obp57a
Genomic Map