Assay Details
| Gene Symbol: Sdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144256.2 | NP_001137728.1 | 2 - 3 | 427 | 71 | |
| NM_001144257.4 | NP_001137729.2 | 2 - 3 | 427 | 71 | ||
| NM_001169767.3 | NP_001163238.2 | 2 - 3 | 427 | 71 | ||
| NM_001369961.1 | NP_001356968.1 | 2 - 3 | 427 | 71 | ||
| NM_057617.5 | NP_476965.1 | 2 - 3 | 427 | 71 | ||
| NM_166449.5 | NP_726071.1 | 2 - 3 | 449 | 71 | ||
| NM_166450.3 | NP_726072.2 | 2 - 3 | 427 | 71 | ||
| GenBank mRNA | AY069377.1 | - | 2 - 3 | 420 | 71 | |
| AY089610.1 | - | 2 - 3 | 424 | 71 | ||
| BT125826.1 | - | 2 - 3 | 387 | 71 | ||
More Information
Set Membership: |
|

Sdc
Genomic Map