Assay Details
| Gene Symbol: HmgZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299765.1 | NP_001286694.1 | 2 - 3 | 368 | 59 | |
| NM_001299766.1 | NP_001286695.1 | 2 - 3 | 447 | 59 | ||
| NM_166476.2 | NP_726106.1 | 2 - 3 | 364 | 59 | ||
| NM_166477.2 | NP_726107.1 | 2 - 3 | 389 | 59 | ||
| GenBank mRNA | AY113439.1 | - | 2 - 3 | 376 | 59 | |
| X71139.1 | - | 375 | 59 | |||
More Information
Set Membership: |
|

HmgZ
Genomic Map