Assay Details
| Gene Symbol: CycB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_166555.3 | NP_726244.1 | 4 - 5 | 1570 | 65 | |
| NM_166556.3 | NP_726245.2 | 4 - 5 | 1564 | 65 | ||
| NM_166557.3 | NP_726246.1 | 4 - 5 | 1493 | 65 | ||
| NM_166558.2 | NP_726247.1 | 3 - 4 | 1485 | 65 | ||
| GenBank mRNA | AY102682.1 | - | 4 - 5 | 1549 | 65 | |
| BT100103.1 | - | 4 - 5 | 1537 | 65 | ||
More Information
Set Membership: |
|

CycB
Genomic Map