Assay Details
| Gene Symbol: stl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103953.3 | NP_001097423.1 | 7 - 8 | 2269 | 61 | |
| NM_001259540.1 | NP_001246469.1 | 7 - 8 | 2777 | 61 | ||
More Information
Set Membership: |
|

stl
Genomic Map