Assay Details
| Gene Symbol: Mthfs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103962.4 | NP_001097432.1 | 1 - 2 | 379 | 105 | |
| NM_001299836.1 | NP_001286765.1 | 1 - 2 | 163 | 105 | ||
More Information
Set Membership: |
|

Mthfs
Genomic Map