Assay Details
Gene Symbol: Acp29AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078788.3 | NP_523512.2 | 1 | 658 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
