Assay Details
| Gene Symbol: tai | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201817.1 | NP_001188746.1 | 11 - 12 | 5804 | 64 | |
| NM_001201819.2 | NP_001188748.1 | 11 - 12 | 5804 | 64 | ||
| NM_001259020.2 | NP_001245949.1 | 9 - 10 | 5999 | 64 | ||
| NM_001273351.1 | NP_001260280.1 | 10 - 11 | 5936 | 64 | ||
| NM_078797.4 | NP_523521.3 | 10 - 11 | 5936 | 64 | ||
More Information
Set Membership: |
|

tai
Genomic Map