Assay Details
| Gene Symbol: Cdk5alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273391.1 | NP_001260320.1 | 2 - 3 | 1347 | 64 | |
| NM_078811.3 | NP_523535.2 | 2 - 3 | 1165 | 64 | ||
| GenBank mRNA | AF231134.1 | - | 2 - 3 | 1115 | 64 | |
More Information
Set Membership: |
|

Cdk5alpha
Genomic Map