Assay Details
| Gene Symbol: Cdk5alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273391.1 | NP_001260320.1 | 4 - 5 | 1695 | 92 | |
| NM_078811.3 | NP_523535.2 | 4 - 5 | 1513 | 92 | ||
More Information
Set Membership: |
|

Cdk5alpha
Genomic Map