Assay Details
| Gene Symbol: Lip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298882.1 | NP_001285811.1 | 1 - 2 | 388 | 69 | |
| NM_078816.4 | NP_523540.1 | 1 - 2 | 388 | 69 | ||
| GenBank mRNA | AY075506.1 | - | 1 - 2 | 302 | 69 | |
More Information
Set Membership: |
|

Lip1
Genomic Map