Assay Details
| Gene Symbol: Lip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298882.1 | NP_001285811.1 | 3 - 4 | 923 | 67 | |
| NM_078816.4 | NP_523540.1 | 3 - 4 | 923 | 67 | ||
| GenBank mRNA | AY075506.1 | - | 3 - 4 | 837 | 67 | |
More Information
Set Membership: |
|

Lip1
Genomic Map