Assay Details
| Gene Symbol: chif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078859.4 | NP_523583.2 | 2 - 3 | 5753 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chif| Gene Symbol: chif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078859.4 | NP_523583.2 | 2 - 3 | 5753 | 59 | |
Set Membership: |
|