Assay Details
| Gene Symbol: Top3alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078878.3 | NP_523602.2 | 1 - 2 | 224 | 64 | |
| GenBank mRNA | BT015282.1 | - | 1 - 2 | 197 | 64 | |
More Information
Set Membership: |
|

Top3alpha
Genomic Map