Assay Details
| Gene Symbol: Itgbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316404.1 | NP_001303333.1 | 4 - 5 | 1203 | 96 | |
| NM_078884.3 | NP_523608.2 | 4 - 5 | 1203 | 96 | ||
| GenBank mRNA | BT031252.1 | - | 4 - 5 | 1188 | 96 | |
More Information
Set Membership: |
|

Itgbn
Genomic Map