Assay Details
| Gene Symbol: Tsp42En | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273829.1 | NP_001260758.1 | 2 - 3 | 420 | 78 | |
| NM_078916.3 | NP_523640.1 | 2 - 3 | 420 | 78 | ||
| GenBank mRNA | AY071666.1 | - | 2 - 3 | 393 | 78 | |
More Information
Set Membership: |
|

Tsp42En
Genomic Map