Assay Details
| Gene Symbol: Tsp42Ep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273830.2 | NP_001260759.1 | 2 - 3 | 247 | 60 | |
| NM_078918.4 | NP_523642.1 | 2 - 3 | 247 | 60 | ||
More Information
Set Membership: |
|

Tsp42Ep
Genomic Map