Assay Details
| Gene Symbol: Tsp42Ep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273830.2 | NP_001260759.1 | 3 - 4 | 603 | 67 | |
| NM_078918.4 | NP_523642.1 | 3 - 4 | 603 | 67 | ||
More Information
Set Membership: |
|

Tsp42Ep
Genomic Map