Assay Details
Gene Symbol: Hey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078933.3 | NP_523657.1 | 1 - 2 | 448 | 67 | |
GenBank mRNA | AF151523.1 | - | 1 - 2 | 743 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
