Assay Details
| Gene Symbol: Socs44A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078935.3 | NP_523659.1 | 1 - 2 | 682 | 71 | |
| GenBank mRNA | AF435923.1 | - | 1 - 2 | 626 | 71 | |
| AY118985.1 | - | 1 - 2 | 626 | 71 | ||
More Information
Set Membership: |
|

Socs44A
Genomic Map