Assay Details
| Gene Symbol: tou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103800.3 | NP_001097270.1 | 4 - 5 | 1386 | 70 | |
| NM_001273969.1 | NP_001260898.1 | 4 - 5 | 1087 | 70 | ||
| NM_001273970.1 | NP_001260899.1 | 5 - 6 | 1352 | 70 | ||
| NM_078977.5 | NP_523701.3 | 4 - 5 | 1386 | 70 | ||
More Information
Set Membership: |
|

tou
Genomic Map