Assay Details
| Gene Symbol: otk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299396.1 | NP_001286325.1 | 4 - 5 | 2154 | 58 | |
| NM_078981.3 | NP_523705.2 | 4 - 5 | 2154 | 58 | ||
| GenBank mRNA | BT015249.1 | - | 4 - 5 | 2240 | 58 | |
More Information
Set Membership: |
|

otk
Genomic Map