Assay Details
| Gene Symbol: otk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078981.3 | NP_523705.2 | 6 - 7 | 3928 | 67 | |
| GenBank mRNA | BT015249.1 | - | 6 - 7 | 4014 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

otk| Gene Symbol: otk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078981.3 | NP_523705.2 | 6 - 7 | 3928 | 67 | |
| GenBank mRNA | BT015249.1 | - | 6 - 7 | 4014 | 67 | |
Set Membership: |
|