Assay Details
| Gene Symbol: Mal-A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136537.3 | NP_610381.1 | 2 - 3 | 877 | 59 | |
| GenBank mRNA | BT031306.1 | - | 2 - 3 | 877 | 59 | |
More Information
Set Membership: |
|

Mal-A4
Genomic Map