Assay Details
| Gene Symbol: Mal-A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169617.2 | NP_001163088.1 | 1 - 2 | 816 | 80 | |
| NM_001299268.1 | NP_001286197.1 | 2 - 3 | 759 | 80 | ||
| NM_136538.4 | NP_610382.2 | 2 - 3 | 759 | 80 | ||
| GenBank mRNA | AY069158.1 | - | 1 - 2 | 767 | 80 | |
| BT016068.1 | - | 2 - 3 | 737 | 80 | ||
More Information
Set Membership: |
|

Mal-A5
Genomic Map