Assay Details
| Gene Symbol: Mal-A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169617.2 | NP_001163088.1 | 3 - 4 | 1727 | 77 | |
| NM_001299268.1 | NP_001286197.1 | 5 - 6 | 1613 | 77 | ||
| NM_136538.4 | NP_610382.2 | 4 - 5 | 1670 | 77 | ||
| GenBank mRNA | AY069158.1 | - | 3 - 4 | 1678 | 77 | |
| BT016068.1 | - | 4 - 5 | 1648 | 77 | ||
More Information
Set Membership: |
|

Mal-A5
Genomic Map