Assay Details
| Gene Symbol: Mal-A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169617.2 | NP_001163088.1 | 4 - 5 | 1870 | 58 | |
| NM_001299268.1 | NP_001286197.1 | 6 - 7 | 1756 | 58 | ||
| NM_136538.4 | NP_610382.2 | 5 - 6 | 1813 | 58 | ||
| GenBank mRNA | AY069158.1 | - | 4 - 5 | 1821 | 58 | |
| BT016068.1 | - | 5 - 6 | 1791 | 58 | ||
More Information
Set Membership: |
|

Mal-A5
Genomic Map