Assay Details
| Gene Symbol: Mal-A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299269.1 | NP_001286198.1 | 2 - 3 | 1732 | 66 | |
| NM_136539.3 | NP_610383.1 | 2 - 3 | 1732 | 66 | ||
More Information
Set Membership: |
|

Mal-A7
Genomic Map