Assay Details
| Gene Symbol: Cirl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144148.4 | NP_001137620.1 | 9 - 10 | 4782 | 62 | |
| NM_001169619.4 | NP_001163090.1 | 9 - 10 | 4770 | 62 | ||
| NM_001259276.2 | NP_001246205.1 | 11 - 12 | 5647 | 62 | ||
| NM_001273878.1 | NP_001260807.1 | 12 - 13 | 5473 | 62 | ||
| NM_136553.5 | NP_610397.4 | 12 - 13 | 5413 | 62 | ||
More Information
Set Membership: |
|

Cirl
Genomic Map