Assay Details
| Gene Symbol: Cirl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144148.4 | NP_001137620.1 | 5 - 6 | 2502 | 82 | |
| NM_001169619.4 | NP_001163090.1 | 5 - 6 | 2490 | 82 | ||
| NM_001259276.2 | NP_001246205.1 | 7 - 8 | 3367 | 82 | ||
| NM_001273878.1 | NP_001260807.1 | 8 - 9 | 3193 | 82 | ||
| NM_136553.5 | NP_610397.4 | 8 - 9 | 3133 | 82 | ||
More Information
Set Membership: |
|

Cirl
Genomic Map