Assay Details
| Gene Symbol: CG14744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103762.3 | NP_001097232.2 | 2 - 3 | 408 | 76 | |
| NM_001299278.1 | NP_001286207.1 | 2 - 3 | 408 | 76 | ||
| NM_001299279.1 | NP_001286208.1 | 2 - 3 | 408 | 76 | ||
More Information
Set Membership: |
|

CG14744
Genomic Map