Assay Details
| Gene Symbol: ana2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136571.3 | NP_610415.1 | 1 - 2 | 850 | 106 | |
| GenBank mRNA | AF132189.1 | - | 1 - 2 | 815 | 106 | |
More Information
Set Membership: |
|

ana2
Genomic Map