Assay Details
| Gene Symbol: MrgBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136573.4 | NP_610417.1 | 1 - 2 | 484 | 57 | |
| GenBank mRNA | AY070912.1 | - | 1 - 2 | 476 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MrgBP| Gene Symbol: MrgBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136573.4 | NP_610417.1 | 1 - 2 | 484 | 57 | |
| GenBank mRNA | AY070912.1 | - | 1 - 2 | 476 | 57 | |
Set Membership: |
|