Assay Details
| Gene Symbol: Mys45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136611.4 | NP_610455.1 | 1 - 2 | 562 | 68 | |
| GenBank mRNA | AF173549.1 | - | 1 - 2 | 482 | 68 | |
| AY070560.1 | - | 1 - 2 | 549 | 68 | ||
More Information
Set Membership: |
|

Mys45A
Genomic Map