Assay Details
| Gene Symbol: Myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273892.1 | NP_001260821.1 | 1 - 2 | 1452 | 101 | |
| NM_001299317.1 | NP_001286246.1 | 1 - 2 | 1118 | 101 | ||
| NM_136635.3 | NP_610479.1 | 1 - 2 | 1452 | 101 | ||
| GenBank mRNA | AF422803.1 | - | 1 - 2 | 1095 | 101 | |
| AY069468.1 | - | 1 - 2 | 1397 | 101 | ||
More Information
Set Membership: |
|

Myd88
Genomic Map