Assay Details
| Gene Symbol: GstT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136665.5 | NP_610509.2 | 3 - 4 | 640 | 84 | |
| GenBank mRNA | AY070580.1 | - | 3 - 4 | 638 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GstT1| Gene Symbol: GstT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136665.5 | NP_610509.2 | 3 - 4 | 640 | 84 | |
| GenBank mRNA | AY070580.1 | - | 3 - 4 | 638 | 84 | |
Set Membership: |
|