Assay Details
| Gene Symbol: Lsm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136678.3 | NP_610522.1 | 1 | 819 | 110 | |
| GenBank mRNA | BT044235.1 | - | 1 | 819 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm11| Gene Symbol: Lsm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136678.3 | NP_610522.1 | 1 | 819 | 110 | |
| GenBank mRNA | BT044235.1 | - | 1 | 819 | 110 | |
Set Membership: |
|